Learn More
Bilobalide, MedChemExpress
MedChemExpress Bilobalide, a sesquiterpene trilactone constituent of Ginkgo biloba, inhibits the NMDA-induced efflux of choline with an IC50 value of 2.3 μM. Bilobalide prevents apoptosis through activation of the PI3K/Akt pathway in SH-SY5Y cells. Exerts protective and trophic effects on neurons.
Brand:  Medchem Express HY-N0076
Encompass_Preferred
 View more versions of this product
Product Code. 16408858
  • 76.95 EUR / 10mg
Description
Bilobalide, a sesquiterpene trilactone constituent of Ginkgo biloba, inhibits the NMDA-induced efflux of choline with an IC50 value of 2.3 μM. Bilobalide prevents apoptosis through activation of the PI3K/Akt pathway in SH-SY5Y cells. Exerts protective and trophic effects on neurons.
Specifications
Specifications
Bilobalide
White
C15H18O8
(-)-Bilobalide
O=C(O1)C[C@@]2([C@]1([H])C[C@]3(C(C)(C)C)O)[C@@]34[C@](OC([C@@H]4O)=O)([H])OC2=O
326.3
Research
Neuroscience-Neurodegeneration
33570-04-6
Research
10 mg
DMSO : ≥ 100 mg/mL (306.47 mM)
326.3
98.0%
Powder