Learn More
Bilobalide, MedChemExpress
MedChemExpress Bilobalide, a sesquiterpene trilactone constituent of Ginkgo biloba, inhibits the NMDA-induced efflux of choline with an IC50 value of 2.3 μM. Bilobalide prevents apoptosis through activation of the PI3K/Akt pathway in SH-SY5Y cells. Exerts protective and trophic effects on neurons.
Brand: Medchem Express HY-N0076
Description
Bilobalide, a sesquiterpene trilactone constituent of Ginkgo biloba, inhibits the NMDA-induced efflux of choline with an IC50 value of 2.3 μM. Bilobalide prevents apoptosis through activation of the PI3K/Akt pathway in SH-SY5Y cells. Exerts protective and trophic effects on neurons.Specifications
Bilobalide | |
White | |
C15H18O8 | |
(-)-Bilobalide | |
O=C(O1)C[C@@]2([C@]1([H])C[C@]3(C(C)(C)C)O)[C@@]34[C@](OC([C@@H]4O)=O)([H])OC2=O | |
326.3 | |
Research | |
Neuroscience-Neurodegeneration |
33570-04-6 | |
Research | |
10 mg | |
DMSO : ≥ 100 mg/mL (306.47 mM) | |
326.3 | |
98.0% | |
Powder |