Learn More
Bilobalide, MedChemExpress
MedChemExpress Bilobalide, a sesquiterpene trilactone constituent of Ginkgo biloba, inhibits the NMDA-induced efflux of choline with an IC50 value of 2.3 μM. Bilobalide prevents apoptosis through activation of the PI3K/Akt pathway in SH-SY5Y cells. Exerts protective and trophic effects on neurons.
76.95 EUR - 429.30 EUR
Specifications
Chemical Name or Material | Bilobalide |
---|---|
CAS | 33570-04-6 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C15H18O8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16404747
|
Medchem Express
HY-N0076 |
50 mg |
245.70 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16418976
|
Medchem Express
HY-N0076 |
100 mg |
429.30 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16408858
|
Medchem Express
HY-N0076 |
10 mg |
76.95 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bilobalide, a sesquiterpene trilactone constituent of Ginkgo biloba, inhibits the NMDA-induced efflux of choline with an IC50 value of 2.3 μM. Bilobalide prevents apoptosis through activation of the PI3K/Akt pathway in SH-SY5Y cells. Exerts protective and trophic effects on neurons.Specifications
Bilobalide | |
White | |
C15H18O8 | |
DMSO : ≥ 100 mg/mL (306.47 mM) | |
326.3 | |
98.0% | |
Powder |
33570-04-6 | |
Research | |
(-)-Bilobalide | |
O=C(O1)C[C@@]2([C@]1([H])C[C@]3(C(C)(C)C)O)[C@@]34[C@](OC([C@@H]4O)=O)([H])OC2=O | |
326.3 | |
Research | |
Neuroscience-Neurodegeneration |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.