Learn More
Lonza™ BioWhittaker™ General-Purpose Serum-Free Media
Chemically defined media optimized for specific cell types
Brand:  Lonza™ 77232
 View more versions of this product
Product Code. 11660121
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
PC-1™
  • Provides complete media system, including PC-1 Supplement for cultivation of primary cells and anchorage-dependent cell lines
  • Contains complete HEPES buffering system
UltraCULTURE™
  • Designed for growth of adherent and nonadherent cells
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2° to 8°C
Serum Free Medium (SFM)
No Glutamine
2 x 500 mL