Learn More
Lonza™ BioWhittaker™ General-Purpose Serum-Free Media
Chemically defined media optimized for specific cell types
683.00 EUR
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
11660121
|
Lonza™
77232 |
Serum Free Medium (SFM) | 2 x 500 mL | ||||||
Description
PC-1™
- Provides complete media system, including PC-1 Supplement for cultivation of primary cells and anchorage-dependent cell lines
- Contains complete HEPES buffering system
- Designed for growth of adherent and nonadherent cells
Specifications
2° to 8°C |