Learn More
Lonza™ BioWhittaker™ RPMI 1640 with L-Glutamine
Developed for culturing normal human leukocytes. BioWhittaker RPMI 1640 with L-glutamine can be used for suspension and monolayer mammalian cultures.
Brand:  Lonza™ 09-774E
 View more versions of this product
Product Code. 11635150
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Manufactured in accordance with cGMP regulations
  • Formulations used for standard liquid media products are those recommended by Tissue Culture Association
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2° to 8°C
RPMI 1640
Sterile-filtered through two consecutive 0.2 μm filters
None
Liquid
Sterile
Glutamine, HEPES, Penicillin, Streptomycin
100 mL