Learn More
Invitrogen™ Human BLC/CXCL13 ELISA Kit
ELISA
706.00 EUR - 5080.00 EUR
Specifications
Accession Number | O43927 |
---|---|
Assay Range | 1.37 to 1000 pg/mL |
Assay Sensitivity | 1.5 pg/mL |
Calibration Range | 1.5 pg/mL to 1000 pg/mL |
Assay | Solid-phase sandwich ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15444963
|
Invitrogen™
EHCXCL13 |
96 Tests |
706.00 EUR
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17175409
|
Invitrogen™
EHCXCL13X5 |
5 x 96 Tests |
2780.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18547814
|
Invitrogen™
EHCXCL13X10 |
10 x 96 Tests |
5080.00 EUR
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Human BLC/CSCL13 (Hu BLC) ELISA quantitates Hu BLC in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu BLC. Principle of the method The Human BLC solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CXCL13, also known as BCA-1 and Angie, is a small chemokine strongly expressed in the secondary lymphoid organs. It preferentially promotes the migration of B lymphocytes (both B-1 and B-2) to the follicles of the spleen, lymph nodes, and Peyer's patches. It is produced by macrophages, and in mouse the highest constitutive producers include large peritoneal macrophages, Kuppfer cells and lung macrophages (both CD11c+ and CD11c-). The cellular ligand for CXCL13 is CXCR5 also known as BLR-1 (Burkitt’s Lymphoma Receptor 1). CXCR5 is expressed on B cells, Tfh cells and, to a lower degree, some memory T cell populations. CXCL13 plays an important role in the development of chronic lymphocytic leukemia (CCL). CXCL13 is also a plasma biomarker for germinal center activity and generation of broadly neutralizing antibodies against HIV.Specifications
O43927 | |
1.5 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
CXCL13 | |
<10 | |
HRP | |
Inflammation | |
2°C to 8°C | |
CXCL13 | |
4 hrs 45 mins |
1.37 to 1000 pg/mL | |
1.5 pg/mL to 1000 pg/mL | |
HRP | |
Growth Factors & Receptors | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL- 12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin, MCP- 1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
10563 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
ANGIE, ANGIE2, BCA-1, BCA1, BLC, BLR1L, SCYB13 | |
1 hrs 20 mins |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.