Learn More
Buerkle™ Blue Detectable Forceps
BIOPHARMAGREENER_CHOICE
Blue detachable forceps feature a special additive in the material that makes them visible to metal detectors or X rays.
Brand:  Buerkle™ 5378-1061
 View more versions of this product
Product Code. 16505220
  • 110.00 EUR / Pack of 100
Estimated Shipment: 22-05-2025
Description
  • Suitable for precise lab work and are ideal for grasping, lifting and moving small objects
  • The fluted handles ensure optimum and secure handling as well as precise work
  • The light plastic forceps for single use allow for fatigue-free work
  • The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
  • The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
  • Individually packaged
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
  • Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
  • Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
  • Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
Specifications
Specifications
Pointed
5.12 in.
130 mm
Sterile
100
Polystyrene
Forceps
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number234191 Certificate TypeCertificate of Compliance Date13/10/2023 Catalog Number5378-1061
Lot Number222962 Certificate TypeCertificate of Compliance Date12/08/2022 Catalog Number5378-1061
Lot Number221707 Certificate TypeCertificate of Compliance Date23/05/2022 Catalog Number5378-1061
Product Title
Buerkle™ Blue Detectable Forceps > Tip Style: Pointed