Learn More
SI Analytics™ BlueLine Plug Cable Combination
SI Analytics™ BlueLine Plug Cable Combination is a coaxial electrode plug that is designed to be used with SI Analytics™ BlueLine series.
Brand:  SI Analytics™ 285122653
 View more versions of this product
Product Code. 10087800
  • 61.40 EUR / Each
Estimated Shipment: 02-05-2025
Description
  • For use with BlueLine series
  • Coaxial cable
  • Available in 1m or 3m lengths
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Coaxial Electrode Plug
LB 1 A
1 m
BlueLine
1m Coaxial Cable, DIN Instrument Plug
Product Title
SI Analytics™ BlueLine Plug Cable Combination > LengthMetricCable: 1m