Learn More
Brady™ BMP71 BMP61 M611 Continuous Matte White Ultra Aggressive Polyester General ID Labels
Ultra Aggressive Polyester (B-489A) with permanent adhesive is ideal for labeling products and general ID in both commercial and industrial applications.
Brand:  Brady™ 011246
 View more versions of this product
Product Code. 11512585
Description
Description
  • Labels for the BMP61 and M611 Thermal Transfer Portable Printers
  • Material is UL & CSA approved with its respective ribbon
  • See the Recommended Ribbon Series for the appropriate ribbon to use
  • Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Specifications
Permanent Rubber Based
Matte
Thermal Transfer
15.24 m x 6.35 mm
White
Polyester (B-489A)
BMP61, BMP71 (with Media Adapter), M611
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ BMP71 BMP61 M611 Continuous Matte White Ultra Aggressive Polyester General ID Labels > 15.24 m x 6.35 mm