Learn More
Berkshire International™ BCR™ Bond Paper
Our BCR™ Bond is made with the purest quality cellulose from specially selected forests and is specially coated to minimize particle and fiber release.
Brand: Berkshire International™ B104A4B25010
30.92 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in 5 colors: Green, Blue, Pink, White, and Yellow
cleanroom, paper, bond
Specifications
Blue | |
Sheets | |
BCR Bond 1040 | |
A4 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
0000846615 | Declaration of Conformity | 06/05/2024 | B104A4B25010 |
0000840311 | Declaration of Conformity | 22/06/2023 | B104A4B25010 |
0000846615 | Certificate of Analysis | 06/05/2024 | B104A4B25010 |
0000840311 | Certificate of Analysis | 22/06/2023 | B104A4B25010 |
0837822 | Certificate of Compliance | 13/12/2021 | B104.A4.B.250/10 |
Product Content Correction
Product Title
Berkshire International™ BCR™ Bond Paper > Page Format: A4, Blue, Bond 1040
Spot an opportunity for improvement?Share a Content Correction