Learn More
Berkshire International™ BCR™ Bond Paper
Our BCR™ Bond is made with the purest quality cellulose from specially selected forests and is specially coated to minimize particle and fiber release.
Brand: Berkshire International™ B68A4BL25010
25.32 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
- Available in 5 colors: Green, Blue, Pink, White, and Yellow
cleanroom, paper, bond
Specifications
Blue | |
Sheets | |
BCR Bond 680 | |
A4 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
0000849583 | Declaration of Conformity | 21/02/2025 | B68A4BL25010 |
0000847356 | Declaration of Conformity | 15/04/2024 | B68A4BL25010 |
0000849583 | Certificate of Analysis | 21/02/2025 | B68A4BL25010 |
0000847356 | Certificate of Analysis | 15/04/2024 | B68A4BL25010 |
0000847356 | Certificate of Compliance | 12/04/2024 | B68A4BL25010 |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
New Offer Save Up to 92%