Learn More
Berkshire International™ BCR™ Bond Paper
BIOPHARMA
Our BCR™ Bond is made with the purest quality cellulose from specially selected forests and is specially coated to minimize particle and fiber release.
Brand:  Berkshire International™ B104A5W25020
15.20 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
 View more versions of this product
Product Code. 12867133
  • 16.00 EUR / Pack of 250
Estimated Shipment: 15-04-2025
Description
  • Available in 5 colors: Green, Blue, Pink, White, and Yellow
cleanroom, paper, bond
Specifications
Specifications
White
Sheets
BCR Bond 1040
A5
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number0000847288 Certificate TypeCertificate of Compliance Date05/03/2024 Catalog NumberB104A5W25020
Lot Number0835516 Certificate TypeCertificate of Compliance Date23/09/2021 Catalog NumberB104.A5.W.250/20
Lot Number0000847288 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberB104A5W25020
Lot Number0000847288 Certificate TypeDeclaration of Conformity Date04/03/2024 Catalog NumberB104A5W25020
Product Title
Berkshire International™ BCR™ Bond Paper > Page Format: A5, White, Bond 1040