missing translation for 'onlineSavingsMsg'
Learn More

Borane-dimethyl sulfide complex, 94%, Thermo Scientific Chemicals

53.16 EUR - 203.01 EUR

Chemical Identifiers

CAS 13292-87-0
Molecular Formula C2H9BS
Molecular Weight (g/mol) 75.96
MDL Number MFCD00013189
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
PubChem CID 9833925
IUPAC Name boron;methylsulfanylmethane
SMILES B.CSC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11446750
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L07705.AC
25 mL
53.16 EUR
25mL
Please to purchase this item. Need a web account? Register with us today!
11456750
View Documents
Thermo Scientific Alfa Aesar
L07705.AE
100 mL
203.01 EUR
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.

Solubility
Miscible with ethyl ether, dichloromethane, benzene, xylene, hexane, diglyme, dimethyl ether, ethyl acetate, toluene, methylene chloride and other aprotic solvents.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Store in a cool place. Incompatible with acids, water, alcohols, bromine, acid anhydrides, acid chlorides and oxidizing agents.
Specifications

Chemical Identifiers

13292-87-0
75.96
RMHDLBZYPISZOI-UHFFFAOYSA-N
9833925
B.CSC
C2H9BS
MFCD00013189
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
boron;methylsulfanylmethane

Specifications

13292-87-0
0.801
−1°C (30°F)
(CH3)2S·BH3
MFCD00013189
3663489
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
RMHDLBZYPISZOI-UHFFFAOYSA-N
boron;methylsulfanylmethane
9833925
94%
-38°C
∼44°C (decomposition)
C2H9BS
1.457
UN2924
Air and moisture sensitive
Miscible with ethyl ether,dichloromethane,benzene,xylene,hexane,diglyme,dimethyl ether,ethyl acetate,toluene,methylene chloride and other aprotic solvents.
B.CSC
75.96
75.97
Borane-dimethyl sulfide complex
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H260-H300-H314-H318-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

P210-P223-P231+P232-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P305+P351+P338-P310-P330-P332+P313-P335+P334-P363-P370+P378q-P501c

H225-H260-H301-H315-H318

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 236-313-6

missing translation for 'rtecsNumber' : PV5080000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold; Store under Argon

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only