missing translation for 'onlineSavingsMsg'
Learn More
Boric Acid, ACS Reagent Grade, Scharlab Product Code.: 11399873
compliance-icons

Boric Acid, ACS Reagent Grade, Scharlab

Product Code. 11399873
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Product Code. 11399873

Brand: Scharlab AC05780500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in building materials, the porcelain industry, cosmetics, the manufacture of dyes, photography and analytical chemistry.

  • ACS Reagent Grade
  • Synonym: Orthoboric acid
  • Molecular Formula: H3BO3
  • Molar Mass: 61.84g/mol
  • CAS: 10043-35-3
  • EIN ECS No.: 233-139-2
  • Organic Matter: Passes test
  • Non-volatile Matter: 0.05% max

Specifications

pH 3.8–4.8 (3.3%, water)
Appearance of Solution Clear and Colorless
CAS 10043-35-3
Melting Point 185°C (decomposition)
Identification Passes Test
Vapor Pressure 2.7hPa at 20°C
Quantity 500 g
Molecular Formula BH3O3
MDL Number MFCD00011337
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
Solubility Information In water: 46.5g/L at 20°C
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES OB(O)O
IUPAC Name boric acid
Molecular Weight (g/mol) 61.83
PubChem CID 7628
ChEBI CHEBI:33118
Formula Weight 61.84g/mol
Percent Purity 99.5 to 100.5% (acidimetric)
Grade ACS Reagent
Show More Show Less
compliance-icons
Product Identifier
  • Boric acid, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boric Acid, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.