missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Description
H3BO3, CAS Number-10043-35-3, 500g, White, 61.83g/mol, 169 deg.C, 11337, Plastic powder jar, 3.8 to 4.8, Powder Solid, 2.7mbar at 20 deg.C

Specifications
Specifications
pH | 3.8 to 4.8 |
Color | White |
Melting Point | 169°C |
Packaging | Plastic powder jar |
Vapor Pressure | 2.7mbar at 20°C |
Quantity | 2.5 kg |
Physical Form | Powder Solid |
Formula Weight | 61.83g/mol |
Safety and Handling

Product Identifier
- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Boric Acid, Certified AR for Analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction