missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Bottom Drip Tray
Material: Stainless Steel; For Use With: UF750TS
Brand: Memmert™ UF750TSB04362
This item is not returnable.
View return policy
Description
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
- Max. loading per tray: 8 kg
Specifications
UF750TS | |
Stainless Steel | |
15 mm rim |
Up to 8 kg | |
Drip Tray |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Bottom Drip Tray > For Use With: UF750TS
Spot an opportunity for improvement?Share a Content Correction