missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Casein Hydrolysate Enzymatic, Powder
BIOPHARMA

Casein hydrolysate enzymatic is recommended for use as Animal diet

115.00 EUR - 464.00 EUR

Specifications

Product Type Animal Diet
Format Powder
For Use With (Application) Animal diet
Storage Requirements Room Temperature
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11463230
BIOPHARMA
View Documents
MP Biomedicals™
0210129001
1 lb.
115.00 EUR
1lb
Please to purchase this item. Need a web account? Register with us today!
11473230
BIOPHARMA
View Documents
MP Biomedicals™
0210129005
5 lb.
464.00 EUR
5lb
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications:
Casein hydrolysate enzymatic is recommended for use as Animal diet.

Product Description:
Casein hydrolysate enzymatic is a pancreatic hydrolysate of casein. This hydrolysate contains, in the form of mixed amino acids and peptides, all amino acids originally present in casein. It is a refined product providing high quality soluble source of amino acids, polypeptides and peptides.

Specifications

Specifications

Animal Diet
Powder
Animal diet
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Casein Hydrolysate Enzymatic, Powder

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.