Learn More
BrandTech™ Slide Staining Kit
VITLAB POM Universal Slide Staining Kit.
Brand:  BrandTech™ 99099
Product Code. 10488036
Description
Description
  • Includes staining jack and staining rack
Specifications
Specifications
Twenty-five 76 x 26 mm slides
One staining jar and one staining rack
100 x 87 x 51 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BrandTech™ Slide Staining Kit > Slide staining kit