Learn More
Micronova™ BucketBinders™ Bucket Connectors
BIOPHARMA
Bucket connecting hardware for BucketBinder™ Multi Bucket System
Brand:  Micronova™ BB-01
283.40 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Product Code. 16235152
  • 301.00 EUR / Each
Estimated Shipment: 23-07-2025
Description
The BucketBinder™ kit includes 2 or 3 polypropylene buckets with casters and one or two sets of BucketBinders (bucket connectors). The system offers easier cleaning and storage - in both small and large controlled environments and the sterile core.
Specifications
Specifications
Bucket Connector, Hardware
Autoclavable
B-7AC Rectangular Bucket
Bucket Connector
Non-sterile
Stainless Steel
Cleanroom Surface Cleaning and Disinfecting
25 cm
1 Pair
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number046585-000 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog NumberBB-01
Lot Number046174-000 Certificate TypeCertificate of Compliance Date01/11/2024 Catalog NumberBB-01
Lot Number045981-000 Certificate TypeCertificate of Compliance Date01/11/2024 Catalog NumberBB-01
Lot Number044984-000 Certificate TypeCertificate of Compliance Date12/08/2024 Catalog NumberBB-01
Lot Number042757-000 Certificate TypeCertificate of Compliance Date14/03/2024 Catalog NumberBB-01
Product Title
Micronova™ BucketBinders™ Bucket Connectors > Bucket connector