Learn More
Thermo Scientific™ Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor
Accessories for the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Brand:  Thermo Scientific™ 75005721
1168.23 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 15241026
  • 1266.30 EUR / Pack of 2
Description
Accessories for the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Max Tube Dimensions: Height <33 mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
M10 rotor
4 Standard or 2 Midi-Deepwell plates
Sealed Bucket
Sealed Bucket; Capacity: 4 Standard or 2 Midi-Deepwell plates (Set of 2)
Certificates
View accessory products
Special Offers
Special Offers
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Get amazing savings on products for your analyses
    Promo Code: 24135
    Expires2025-07-13
Product Content Correction
Product Title
Thermo Scientific™ Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor >