Learn More
Buffer Colour Coded Concentrated Solution pH4.00 (Phthalate) Red, NIST Standard for pH measurement, Fisher Chemical™
70.34 EUR - 377.34 EUR
Specifications
MDL Number 217672
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10164863
View Documents
Fisher Chemical
J/2825C/05
1 Ampule This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10090200
View Documents
Fisher Chemical
J/2825C/90
6 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Specifications
217672
Documents
Product Certifications
Product Content Correction
Product Title
Buffer Colour Coded Concentrated Solution pH4.00 (Phthalate) Red, NIST Standard for pH measurement, Fisher Chemical™