missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP130 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA586277
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a signal transducer shared by many cytokines, including interleukin 6 (IL6), ciliary neurotrophic factor (CNTF), leukemia inhibitory factor (LIF), and oncostatin M (OSM). This protein functions as a part of the cytokine receptor complex. The activation of this protein is dependent upon the binding of cytokines to their receptors. vIL6, a protein related to IL6 and encoded by the Kaposi sarcoma-associated herpesvirus, can bypass the interleukin 6 receptor (IL6R) and directly activate this protein. Knockout studies in mice suggested a critical role of the gene encoding this protein in regulating myocyte apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
GP130 | |
Polyclonal | |
Unconjugated | |
IL6ST | |
5133400A03Rik, AA389424, Ac1055, BB405851, CD130, CD130 antigen, CDW130, D13Ertd699e, EGK_16499, Gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, gp130, oncostatin M receptor, GP130-RAPS, IL-6, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6R-beta, IL6ST, interleukin 6 signal transducer, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin 6 signal transducer receptor, interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, interleukin-6 signal transducer receptor, membrane glycoprotein, membrane glycoprotein 130, membrane glycoprotein gp130, oncostatin-M receptor subunit alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16195, 3572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide, pH 7.2 | |
P40189, Q00560 | |
IL6ST | |
Synthetic peptide, corresponding to amino acids 750-800 of Human gp130. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GP130 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction