missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C8orf33 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18500AF532
This item is not returnable.
View return policy
Description
C8orf33 Polyclonal antibody specifically detects C8orf33 in Human samples. It is validated for Western Blot
Specifications
C8orf33 | |
Polyclonal | |
Western Blot | |
chromosome 8 open reading frame 33, FLJ20989, hypothetical protein LOC65265 | |
Recognizes region between residues Between between 25 and 75.. | |
0.1 mL | |
Stem Cell Markers | |
65265 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C8orf33 Antibody, Alexa Fluor™ 532, Novus Biologicals™ > 0.1 mL; Alexa Fluor 532
Spot an opportunity for improvement?Share a Content Correction