Learn More
Calcium D-saccharate tetrahydrate, 98%, Thermo Scientific Chemicals
89.55 EUR - 382.91 EUR
Chemical Identifiers
CAS | 5793-89-5 |
---|---|
Molecular Formula | C6H16CaO12 |
Molecular Weight (g/mol) | 320.26 |
MDL Number | MFCD00150704 |
InChI Key | ZQWFSIZRQANUDA-WQMSYZFBSA-L |
Synonym | calcium saccharate, unii-6ap9j91k4v, d-saccharic acid calcium salt tetrahydrate, calcium d-glucarate tetrahydrate, calcium d-saccharate tetrahydrate, calcium saccharat-4-wasser, calcium saccharate usan:usp:inn, calcium saccharate usp, calcium d-glucarate, tetrahydrate, calcium d-glucarate 1:1 , tetrahydrateShow More |
PubChem CID | 16211545 |
IUPAC Name | calcium;(2R,3S,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate;tetrahydrate |
SMILES | C(C(C(C(=O)[O-])O)O)(C(C(=O)[O-])O)O.O.O.O.O.[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438519
|
Thermo Scientific Alfa Aesar
J66947.14 |
25 g |
89.55 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15448519
|
Thermo Scientific Alfa Aesar
J66947.22 |
100 g |
382.91 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.
Solubility
Soluble in water(0.43g/L).
Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
5793-89-5 | |
320.26 | |
ZQWFSIZRQANUDA-WQMSYZFBSA-L | |
16211545 | |
C(C(C(C(=O)[O-])O)O)(C(C(=O)[O-])O)O.O.O.O.O.[Ca+2] |
C6H16CaO12 | |
MFCD00150704 | |
calcium saccharate, unii-6ap9j91k4v, d-saccharic acid calcium salt tetrahydrate, calcium d-glucarate tetrahydrate, calcium d-saccharate tetrahydrate, calcium saccharat-4-wasser, calcium saccharate usan:usp:inn, calcium saccharate usp, calcium d-glucarate, tetrahydrate, calcium d-glucarate 1:1 , tetrahydrateShow More | |
calcium;(2R,3S,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate;tetrahydrate |
Specifications
5793-89-5 | |
Powder | |
MFCD00150704 | |
14,1700 | |
Soluble in water(0.43g/L). | |
C(C(C(C(=O)[O-])O)O)(C(C(=O)[O-])O)O.O.O.O.O.[Ca+2] | |
320.26 | |
320.26 | |
Odorless |
White | |
C6H16CaO12 | |
3821334 | |
calcium saccharate, unii-6ap9j91k4v, d-saccharic acid calcium salt tetrahydrate, calcium d-glucarate tetrahydrate, calcium d-saccharate tetrahydrate, calcium saccharat-4-wasser, calcium saccharate usan:usp:inn, calcium saccharate usp, calcium d-glucarate, tetrahydrate, calcium d-glucarate 1:1 , tetrahydrate | |
ZQWFSIZRQANUDA-WQMSYZFBSA-L | |
calcium;(2R,3S,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate;tetrahydrate | |
16211545 | |
98% | |
Calcium D-saccharate tetrahydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 227-334-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.