Learn More
WTW™ Calibration and Maintenance Set
Calibration and maintenance are complete assortments for the calibration and maintenance of electrodes. The pepsin cleaning solution is used to remove protein containing precipitation from diaphragms, the 3 molar KCI as a storage or refill solution.
85.10 EUR - 119.00 EUR
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
10398683
|
WTW™
108825 |
pH-3 Technical buffer solution 250mL (pH 4.01, pH 7, pH 10.01) trace, 2 bottles of 250mL of KCl 3mol/L bottles |
85.10 EUR
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
10634471
|
WTW™
109415 |
With standard (DIN/NBS) buffer solution, 3 bottles, 250mL each of pH 4.006 - 6.865 - 9.180, 1 bottle with 250mL pepsin cleaning solution, 1 bottle with 250mL KCl solution 3M/L |
119.00 EUR
1 set |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
Description
- Adapted to different electrode types
- Cost-effective combinations
- Calibration and maintenance in one assortment
Specifications
Calibration and Maintenance Set |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
WTW™ Calibration and Maintenance Set