Learn More
CAPN1 293T Cell Overexpression Lysate (Denatured), Abnova
Brand:  Abnova H00000823-T01.100uL
Product Code. 16145604
  • 215.00 EUR / 100µL
Description
Western Blotting
Specifications
Specifications
293T
CAPN1 293T Cell Transient Overexpression Lysate(Denatured)
78.65
pCMV-CAPN1 full-length
100 μL
Human
Western Blot
NM_005186
6.8
Transient overexpression cell Lysate was tested with Anti-CAPN1 antibody (H00000823-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
Product Title
CAPN1 293T Cell Overexpression Lysate (Denatured), Abnova > 100μL