Learn More
Thermo Scientific™ CaptureSelect™ Human Fab-lambda Kinetics Biotin Conjugate
Thermo Scientific™ CaptureSelect™ Human Fab-lambda Kinetics Biotin Conjugate is a bi-functional ligand construct consisting of 2 different 13 kDa recombinant single domain antibody fragments (affinity ligands).
543.00 EUR - 1900.00 EUR
Specifications
Content And Storage | Store in freezer (-5°C to -30°C). |
---|---|
Form | Liquid |
Product Line | CaptureSelect™ |
Type | Conjugate |
Label or Dye | Biotin & Other Haptens |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
18496818
|
Thermo Scientific™
7103312100 |
100 μg |
543.00 EUR
100µg |
In Stock
Log in to see stock. |
||||||
18426678
|
Thermo Scientific™
7103312500 |
500 μg |
1900.00 EUR
500µg |
In Stock
Log in to see stock. |
||||||
Description
CaptureSelect Human Fab-lambda Kinetics Biotin Conjugate is a bi-functional ligand construct consisting of 2 different 13 kDa recombinant single domain antibody fragments (affinity ligands).
The CaptureSelect anti-LC-lambda (Hu) ligand that specifically binds to the constant domain of human lambda light chains is genetically fused to the CaptureSelect anti-IgG-CH1 ligand that specifically binds to the CH1 domain of all four human IgG subclasses. The bi-functional affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect Human Fab-lambda Kinetics Biotin Conjugate allows you to:
- Screen Fab-antigen interactions—the Human Fab-lambda Kinetics Biotin Conjugate allows high affinity capture of human IgG Fab fragments that possess a lambda light chain. Extreme low off-rates [k(diss)] are achieved through avidity by targeting the CL-lambda domain of the Fab (light chain) and its IgG CH1 domain (heavy chain) simultaneously, thereby enabling Fab screening on target binding kinetics.
Applications for CaptureSelect Human Fab-lambda Kinetics Biotin Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
Store in freezer (-5°C to -30°C). | |
CaptureSelect™ | |
Biotin & Other Haptens |
Liquid | |
Conjugate | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.