missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1 Antibody (76-7-4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP1-28221
This item is not returnable.
View return policy
Description
CD1 Monoclonal specifically detects CD1 in Porcine samples. It is validated for Flow Cytometry.
Specifications
CD1 | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000 | |
CD1A | |
Fresh dd miniature swine thymocytes | |
0.25 mg | |
Primary | |
Porcine CD1 | |
Store at 4C. Do not freeze. | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunocytochemistry | |
76-7-4 | |
Unconjugated | |
CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
Mouse | |
Protein A or G purified | |
RUO | |
909 | |
Pig | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD1 Antibody (76-7-4) - Azide and BSA Free, Novus Biologicals™ > 0.25mg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction