missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Monoclonal Antibody (N418), Brilliant Violet™ 711, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
185.00 EUR - 379.00 EUR
Specifications
Antigen | CD11c |
---|---|
Clone | N418 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019755
|
Invitrogen
407-0114-82 |
100μg |
379.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17825067
|
Invitrogen
407-0114-80 |
25 μg |
185.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.Specifications
CD11c | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q9QXH4 | |
16411 | |
IgG | |
Affinity chromatography | |
Itgax |
N418 | |
Flow Cytometry | |
Brilliant Violet 711 | |
Armenian Hamster | |
Mouse | |
AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6 | |
Itgax | |
Primary | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD11c Monoclonal Antibody (N418), Brilliant Violet™ 711, eBioscience™, Invitrogen™