Learn More
CD31 Rabbit anti-Human, Unconjugated, Clone: 4N13, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80530-1-RR-100UL
This item is not returnable.
View return policy
Description
The CD31 antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human CD31. This antibody recognizes human antigen. The CD31 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CD31 | |
4N13 | |
Rabbit | |
Primary | |
Human |
Western Blot | |
Unconjugated | |
100 μL | |
5175 | |
IgG |