missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31 Rabbit anti-Human, Unconjugated, Clone: 4N13, Proteintech
Rabbit Recombinant Antibody
149.00 EUR - 439.00 EUR
Specifications
Antigen | CD31 |
---|---|
Clone | 4N13 |
Applications | Western Blot |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17591506
|
Proteintech
80530-1-RR-20UL |
20 μL |
149.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17581506
|
Proteintech
80530-1-RR-100UL |
100 μL |
439.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD31 antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human CD31. This antibody recognizes human antigen. The CD31 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CD31 | |
Western Blot | |
Rabbit | |
5175 | |
Primary |
4N13 | |
Unconjugated | |
Human | |
IgG |