missing translation for 'onlineSavingsMsg'
Learn More

CD8 alpha Monoclonal Antibody (5H10), Pacific Blue™, Invitrogen™

Rat Monoclonal Antibody

167.00 EUR - 404.00 EUR

Specifications

Antigen CD8 alpha
Clone 5H10
Applications Flow Cytometry
Classification Monoclonal
Conjugate Pacific Blue
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10641254
missing translation for 'viewDocuments'
Invitrogen
MCD0828
1 mL
404.00 EUR
Each
Please to purchase this item. Need a web account? Register with us today!
11360772
missing translation for 'viewDocuments'
Invitrogen
MCD0828TR
25 Tests
167.00 EUR
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cluster of differentiation 8 (CD8), a type I transmembrane glycoprotein of the immunoglobulin family of receptors, plays an integral role in signal transduction, and T cell differentiation and activation. CD8 is predominantly expressed on T cells as a disulfide-linked heterodimer of CD8alpha and CD8beta, where it functions as a co-receptor, along with T cell receptor (TCR), for major histocompatibilty complex class I (MHC-I) molecules; whereas its counterpart, CD4, acts as a co-receptor for MHC-II molecules. CD8 exists on the cell surface, where the CD8alpha chain is essential for binding to MHC-I. CD8 is also expressed on a subset of T cells, NK cells, monocytes and dendritic cells as disulfide-linked homodimers of CD8alpha. Ligation of MHC-I/peptide complexes presented by antigen-presenting cells (APCs), triggers the recruitment of lymphocyte-specific protein tyrosine kinase (Lck), which leads to lymphokine production, motility and cytotoxic T lymphocyte (CTL) activation. Once activated, CTLs play a crucial role in the clearance of pathogens and tumor cells. Differentiation of naive CD8+ T cells into CTLs is strongly enhanced by IL-2, IL-12 and TGF-beta1.
Specifications
Show More
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD8 alpha Monoclonal Antibody (5H10), Pacific Blue™, Invitrogen™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.