missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC16 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77155AF647
This item is not returnable.
View return policy
Description
CDC16 Polyclonal antibody specifically detects CDC16 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
CDC16 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
ANAPC6subunit 6, CDC16 (cell division cycle 16, S. cerevisiae, homolog), CDC16 cell division cycle 16 homolog (S. cerevisiae), CDC16 homolog, CDC16Hs, cell division cycle 16 homolog (S. cerevisiae), cell division cycle protein 16 homolog, Cyclosome subunit 6 | |
Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human APC6. The immunogen is located within the first 50 amino acids of APC6. Amino Acid Squence: DKVASLSREEPQDIYWLA | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Ubiquitin Proteasome Pathway | |
8881 | |
Store at 4°C in the dark. | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CDC16 Antibody, Alexa Fluor™ 647, Novus Biologicals™ > 0.1 mL; Alexa Fluor 647
Spot an opportunity for improvement?Share a Content Correction