missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CDC42 BPG (MRCKG), GST Tag Recombinant Protein
Human CDC42 BPG (MRCKG), GST Tag Recombinant Protein
Brand: Invitrogen™ A30490
This item is not returnable.
View return policy
Description
- This CDC42 BPG (MRCKG) recombinant human protein (1–475) was expressed in insect cells
- CDC42 BPG serine/threonine kinase is encoded by the CDC42BPG gene
- It acts as a downstream effector of CDC42 in cytoskeletal reorganization, and contributes to the actomyosin contractility required for cell invasion
- It belongs to the AGC protein kinase family, DMPK subfamily
- CDC42 BPG (MRCKG) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the Z'-LYTE™ assay
- Available in multiple pack sizes for convenient use
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
NP_059995 | |
80.2 kDa | |
Other Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
10 μg | |
aa 1 - 475 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Baculovirus | |
Recombinant, Active, Catalytic |