missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Cell Line Optimization 4D-Nucleofector™ X Kit
Brand: Lonza™ V4XC-9064
This item is not returnable.
View return policy
Description

Specifications
4°C | |
Transfection efficiency up to 84% . | |
plasmid DNA and/or siRNA | |
64 | |
1 Each |
pmaxGFP™ | |
4D-Nucleofector™ X Unit | |
Includes 20μL Transfection volume, 0.675mL SE, SF and SG 4D-Nucleofector X Solution, 3 x 0.15mL Supplement, 50μg pmaxGFP Vector, 4 x 20μL 16-well Nucleocuvette Strips | |
Cell Line Optimization 4D-Nucleofector X Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lonza™ Cell Line Optimization 4D-Nucleofector™ X Kit > Number of Reactions: 64
Spot an opportunity for improvement?Share a Content Correction