missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CellVation™ Cryopreservation Medium

Recommended for the preservation of stem cells, hepatocytes, tissue samples, and a wide variety of cell types and established cell lines

335.00 EUR

Specifications

Certifications/Compliance Mycoplasma, Bacteria, Fungi, Yeast and Mold free. Certified U.S. origin.
Content And Storage Store at 4°C
Description CellVation Cryopreservation Media
Form Solution
Product Type Cryopreservation Media
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11454855
View Documents
MP Biomedicals™
0920300M2
60 mL
335.00 EUR
60mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CellVation is a cryopreservation medium designed for optimal cell recovery and convenience. It does not contain DMSO or serum, and is scientifically engineered for serum-free cultures, as well as, ideally suited for all types of serum. It has also been shown to preserve the potential of stem cells for differentiation.

Specifications

Specifications

Mycoplasma, Bacteria, Fungi, Yeast and Mold free. Certified U.S. origin.
CellVation Cryopreservation Media
Cryopreservation Media
Cell counting, viability and cryopreservation
Store at 4°C
Solution
<0.5 EU/mL
6.5 to 7.5
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.