Learn More
Eppendorf™ Centrifuge Adapters For Rotors for 5810 and 5810R
Available in a variety of configurations to suit a wide range of applications
Brand: Eppendorf™ 5810752000
182.99 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
This item is not returnable.
View return policy
Description
- Adaptors for Eppendorf™ Centrifuges 5810 and 5810R.
- Compatible with conical or flat tubes
Specifications
Adapter | |
2.6 to 7 mL | |
13 mm | |
2 Pack |
Flat | |
Rotors 5810 and 5810R Centrifuge | |
60 |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Product Content Correction
Product Title
Eppendorf™ Centrifuge Adapters For Rotors for 5810 and 5810R > Capacity: 2.6 to 7mL; Number of Places: 60
Spot an opportunity for improvement?Share a Content Correction