missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP290 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539088
This item is not returnable.
View return policy
Description
CEP290 activates ATF4 mediated transcription and is required for the correct localization of ciliary and phototransduction proteins in retinal photoreceptor cells; may play a role in ciliary transport processes. CEP290 is ubiquitously expressed; strongly in placenta and weakly in brain. There are two named isoforms. Defects in CEP290 are a cause of Joubert syndrome type 5 (JBTS5), Senior-Loken syndrome type 6 (SLSN6), Leber congenital amaurosis type 10 (LCA10) and Meckel syndrome type 4 (MKS4). Antibodies against CEP290 are present in sera from patients with cutaneous T cell lymphomas, but not in the healthy control population.
Specifications
CEP290 | |
Polyclonal | |
Unconjugated | |
CEP290 | |
Centrosomal protein of 290 kDa, Cep290, Bardet-Biedl syndrome 14 protein, Cancer/testis antigen 87, CT87, Nephrocystin-6, Tumor antigen se2-2, CEP290, BBS14, KIAA0373, NPHP6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
216274, 80184 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15078, Q6A078 | |
CEP290 | |
A synthetic peptide derived from the internal region of human CEP290 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CEP290 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction