Learn More
CF53, MedChemExpress
MedChemExpress CF53 is a highly potent, selective and orally active inhibitor of BET protein, with a Ki of Kd of 2.2 nM and an IC50 of 2 nM for BRD4 BD1. CF53 binds to both the BD1 and BD2 domains of BRD2, BRD3, BRD4, and BRDT BET proteins with high affinities, very selective over non-BET bromodomain-containing proteins. CF53 shows potent anti-tumor activity both in vitro and in vivo.

515.70 EUR - 2301.75 EUR
Specifications
Chemical Name or Material | CF53 |
---|---|
CAS | 1808160-52-2 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C24H25N7O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16470947
![]() |
Medchem Express
HY-112610 |
50 mg |
1701.00 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16464658
![]() |
Medchem Express
HY-112610 |
10 mg |
773.55 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16437377
![]() |
Medchem Express
HY-112610 |
5 mg |
515.70 EUR
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16435496
![]() |
Medchem Express
HY-112610 |
100 mg |
2301.75 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CF53 is a highly potent, selective and orally active inhibitor of BET protein, with a Ki of Kd of 2.2 nM and an IC50 of 2 nM for BRD4 BD1. CF53 binds to both the BD1 and BD2 domains of BRD2, BRD3, BRD4, and BRDT BET proteins with high affinities, very selective over non-BET bromodomain-containing proteins. CF53 shows potent anti-tumor activity both in vitro and in vivo.Specifications
CF53 | |
Light Yellow | |
C24H25N7O2 | |
COC1=C(C2=C(C)ON=C2C)C=C(NC3=C4C(NC5=CC(C6CC6)=NN5C)=NC(C)=N3)C4=C1 | |
443.5 | |
Research | |
Cancer-programmed cell death |
1808160-52-2 | |
Research | |
DMSO : 110 mg/mL (248.03 mM; Need ultrasonic) | |
443.5 | |
98.09% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : -20°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Recommended Storage : -20°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.