Learn More
CGP 25454A, MedChemExpress
MedChemExpress CGP 25454A is a novel and selective presynaptic dopamine autoreceptor antagonist.
Brand: Medchem Express HY-100454

Description
CGP 25454A is a novel and selective presynaptic dopamine autoreceptor antagonist.Specifications
CGP 25454A | |
Off-White | |
C15H21Cl2N3O2 | |
DMSO : ≥ 3.5 mg/mL (10.11 mM) | |
346.25 | |
99.46% | |
Powder |
104391-26-6 | |
Research | |
50 mg | |
O=C(NCCN(CC)CC)C1=CC(C#N)=C(Cl)C=C1OC.[H]Cl | |
346.25 | |
Research | |
Neuroscience-Neuromodulation |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only