Learn More
Eppendorf™ Charger Stand 2, for Eppendorf Xplorer™/Xplorer™ plus
Saves bench space and conveniently stores and charges your pipette.
Brand: Eppendorf™ 3116000031
281.99 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Saves bench space and conveniently stores and charges your pipette.
- Carries and charges one Eppendorf Xplorer™ or Xplorer™ plus pipette
- Robust design that securely holds and protects the instrument
- Large rubber feet protect stand from liquids spilled on workbench
- Cord wrap functionality for unused power cord
- Requires mains/power adapter (not included); power adapter is supplied with Eppendorf Xplorer™ or Xplorer™ plus Pipette
Specifications
Charger Stand 2 | |
Pipette Holder and Charger | |
One Eppendorf Xplorer™ or Xplorer plus pipette | |
Storing and charging pipettes |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%