Learn More
Palintest™ Chlorine DPD 1 & 3 Photometer Reagents
Test for chlorine using these tablets that rely on the DPD method developed by Dr. A. T. Palin. Palintest™ Chlorine DPD 1 & 3 Photometer Reagents are very convenient and easy to use.
Brand:  Palintest™ PM031
Product Code. 11924666
  • 31.00 EUR / Pack of 50
Description
  • DPD method - standard method for testing for chlorine and other disinfectant residuals.
  • Tablet form for maximum convenience and simplicity of use.
  • Measures free chlorine concentration and total chlorine concentration.
  • Measure color intensities with a Palintest™ photometer.
Specifications
Specifications
50
0 to 5mg/L
Chlorine DPD 1 & 3 Test Reagent Tablets
50/Pk.
50 Tests
Product Title
Palintest™ Chlorine DPD 1 & 3 Photometer Reagents > Range: 0 to 5mg/L