missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Cholic acid, sodium salt, 98+%, Thermo Scientific Chemicals Product Code.: 10321061

Thermo Scientific Chemicals Cholic acid, sodium salt, 98+%, Thermo Scientific Chemicals

Product Code. 10321061
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10321061

Brand: Thermo Scientific Chemicals 405191000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cholic acid, sodium salt, CAS # 361-09-1, is a bile acid that can be non-denaturing detergent and can be used for membrane protein extraction.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Cholic acid, sodium salt, is a bile acid that can aid digestion of fats and certain nutrients

Application
• It can facilitate digestion of lipids
• It is a non-denaturing detergent
• It can be used in combination with urea for extraction of membrane protein and protein complexes

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 361-09-1
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.56
MDL Number MFCD00150749,MFCD00064138
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
Synonym sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, sodium cholate, reagent, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt
PubChem CID 57349315
IUPAC Name sodium;(4R)-4-[(8R,9S,10S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Color White
pH 7.5 to 9.5 (5% aq. soln.)
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Merck Index 14, 2203
Formula Weight 430.55
Percent Purity 98.0 to 102.0% (on dried basis)
Infrared Spectrum Authentic
Loss on Drying 5.0% max. (110°C)
Packaging Glass bottle
Chemical Name or Material Cholic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Cholic acid, sodium salt, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.