Learn More
Macherey-Nagel™ Chromabond™ Adapter
Brand:  Macherey-Nagel™ 730100.4
 View more versions of this product
Product Code. 12416546
Specifications
Combination with 1, 3, 6 mL Polypropylene Column
Adapter
4 Pack
Product Title
Macherey-Nagel™ Chromabond™ Adapter > For Use With: For Combination with 1, 3, 6mL Polypropylene Column