Learn More
Macherey-Nagel™ Chromafix™ PS-Ag+ SPE Cartridge
Brand: Macherey-Nagel™ 731866
282.54 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
PS/DVB Copolymer | |
RP and Ion Chromatography | |
Chromafix PS-Ag+ | |
100 Å |
SPE | |
100 μm | |
SPE Cartridge |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Macherey-Nagel™ Chromafix™ PS-Ag+ SPE Cartridge > Adsorbent Weight: 240mg
Spot an opportunity for improvement?Share a Content Correction