Learn More
Scharlab CHROMagar™ Candida
CHROMagar Candida is a chromogenic dehydrated medium for the isolation and differentiation of major clinical-significant Candida species.
Brand: Scharlab 00000CA222
This item is not returnable.
View return policy
Description
- Chromogenic medium for the detection of Candida spp.
- More commonly, Candida species are involved in superficial oropharyngeal and urogenital infections.
- Although C.albicans remains the major species involved, other types such as C.tropicalis, C.krusei or C.glabrata have increased proportionally as new antifungal agents have worked very effectively against C.albicans.
Specifications
Powder | |
Differential Medium | |
Isolation and Differentiation of Major Clinical-Significant Candida Species | |
48 hr. | |
5 L |
CHROMagar™ Candida | |
Agar 15.0 g/L, Peptone 10.2 g/L, Chloramphenicol 0.5 g/L, Chromogenic Mix 22.0 g/L | |
30°C to 37°C | |
6.1 ±0.2 |