missing translation for 'onlineSavingsMsg'
Learn More

Scharlab CHROMagar™ Candida

CHROMagar Candida is a chromogenic dehydrated medium for the isolation and differentiation of major clinical-significant Candida species.

774.00 EUR - 3210.00 EUR

Specifications

Media Type CHROMagar™ Candida
Product Type Differential Medium
Formulation Agar 15.0 g/L, Peptone 10.2 g/L, Chloramphenicol 0.5 g/L, Chromogenic Mix 22.0 g/L
For Use With (Application) Isolation and Differentiation of Major Clinical-Significant Candida Species
Incubation Conditions 30°C to 37°C
View More Specs

Products 2
Product Code Brand Yield Price Quantity & Availability  
Product Code Brand Yield Price Quantity & Availability  
11406277
View Documents
Scharlab
00000CA222
5 L
774.00 EUR
5 L
Estimated Shipment: 07-03-2025
to see stock.
Add to basket
 
11416277
View Documents
Scharlab
00000CA223
25 L
3210.00 EUR
25 L
Estimated Shipment: 07-03-2025
to see stock.
Add to basket
 
Description

Description

  • Chromogenic medium for the detection of Candida spp.
  • More commonly, Candida species are involved in superficial oropharyngeal and urogenital infections.
  • Although C.albicans remains the major species involved, other types such as C.tropicalis, C.krusei or C.glabrata have increased proportionally as new antifungal agents have worked very effectively against C.albicans.
Specifications

Specifications

CHROMagar™ Candida
Agar 15.0 g/L, Peptone 10.2 g/L, Chloramphenicol 0.5 g/L, Chromogenic Mix 22.0 g/L
30°C to 37°C
6.1 ±0.2
Differential Medium
Isolation and Differentiation of Major Clinical-Significant Candida Species
48 hr.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.