Learn More
Macherey-Nagel™ Chrombond™ RS 40 SiOH Polypropylene Flash Column
Brand: Macherey-Nagel™ 732803
This item is not returnable.
View return policy
Specifications
171 mm | |
Flash Column | |
Directly Used in The Teledyne Isco Companion, Rf |
Polypropylene | |
Flash Column |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Macherey-Nagel™ Chrombond™ RS 40 SiOH Polypropylene Flash Column > I.D.: 26.4mm
Spot an opportunity for improvement?Share a Content Correction