missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED1 Monoclonal Antibody (OTI1C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807035S
This item is not returnable.
View return policy
Description
CITED1 Monoclonal antibody specifically detects CITED1 in Human samples. It is validated for Western BlotSpecifications
CITED1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8TDQ0 | |
Mouse | |
Affinity chromatography | |
RUO | |
84868 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Western Blot | |
OTI1C6 | |
Unconjugated | |
Havcr2 | |
AI316840; AU019144; Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1; cbp/p300-interacting transactivator 1; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 1; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1; Cited1; CITED1 protein; melanocyte-specific gene 1; melanocyte-specific gene 1 protein; melanocyte-specific protein 1; melanocyte-specific protein 1; p35srj; CBP/P300-interacting transactivator 1; CITED1; Msg1; MSG1 protein; transactivator, with Glu/Asp-rich carboxy-terminal domain, 1 | |
Human recombinant protein fragment corresponding to amino acids 22-202 of human HAVCR2 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |