missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED1 Monoclonal Antibody (OTI1C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
242.00 EUR - 540.87 EUR
Specifications
Antigen | CITED1 |
---|---|
Clone | OTI1C6 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17507383
|
Origene Technologies
TA807035S |
30 μL |
242.00 EUR
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17587373
|
Origene Technologies
TA807035 |
100 μL |
540.87 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CITED1 Monoclonal antibody specifically detects CITED1 in Human samples. It is validated for Western BlotSpecifications
CITED1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
IgG2b | |
-20° C, Avoid Freeze/Thaw Cycles |
OTI1C6 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Primary |